Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPTX1
Entrez Gene ID
513538
Gene Name
neuronal pentraxin 1
Gene Aliases
-
Chromosome Location
Chr.19: 52059956 - 52066350 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192354.1 NP_001179283.1 1 - 2 585 59

View Full Product Details