Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STOML1
Entrez Gene ID
540438
Gene Name
stomatin like 1
Gene Aliases
-
Chromosome Location
Chr.21: 34633172 - 34648683 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192431.1 NP_001179360.1 2 - 3 252 61

View Full Product Details