Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KBTBD12
Entrez Gene ID
516527
Gene Name
kelch repeat and BTB domain containing 12
Gene Aliases
KLHDC6
Chromosome Location
Chr.22: 59577852 - 59628254 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192448.3 NP_001179377.1 2 - 3 1342 83

View Full Product Details