Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED8
Entrez Gene ID
614106
Gene Name
transmembrane p24 trafficking protein family member 8
Gene Aliases
-
Chromosome Location
Chr.10: 88544185 - 88581770 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193012.1 NP_001179941.1 1 - 2 121 71

View Full Product Details