The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193081.2 | NP_001180010.2 | 2 - 3 | 244 | 70 |
GenBank mRNA | CK776230.1 | - | 2 - 3 | 244 | 70 |