Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTDSPL
Entrez Gene ID
538425
Gene Name
CTD small phosphatase like
Gene Aliases
-
Chromosome Location
Chr.22: 11308887 - 11433803 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193081.2 NP_001180010.2 6 - 7 534 105
GenBank mRNA CK776230.1 - 6 - 7 534 105

View Full Product Details