Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VSIG2
Entrez Gene ID
616943
Gene Name
V-set and immunoglobulin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.29: 28226459 - 28231686 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193134.2 NP_001180063.2 1 - 2 60 107

View Full Product Details