Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLRN2
Entrez Gene ID
617290
Gene Name
clarin 2
Gene Aliases
-
Chromosome Location
Chr.6: 112942655 - 112952422 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193151.1 NP_001180080.1 1 - 2 253 95

View Full Product Details