Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM47
Entrez Gene ID
618638
Gene Name
tripartite motif containing 47
Gene Aliases
-
Chromosome Location
Chr.19: 55741289 - 55745792 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193197.3 NP_001180126.3 4 - 5 1238 74

View Full Product Details