Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX11G
Entrez Gene ID
506518
Gene Name
peroxisomal biogenesis factor 11 gamma
Gene Aliases
-
Chromosome Location
Chr.7: 16347584 - 16372485 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193256.1 NP_001180185.1 2 - 3 287 91

View Full Product Details