Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAMSTR
Entrez Gene ID
505540
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
-
Chromosome Location
Chr.18: 55352451 - 55358248 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195011.1 NP_001181940.1 1 - 2 55 62

View Full Product Details