Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAMSTR
Entrez Gene ID
505540
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
-
Chromosome Location
Chr.18: 55352451 - 55358248 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195011.1 NP_001181940.1 5 - 6 557 108
GenBank mRNA BC151712.1 - 4 - 5 500 108

View Full Product Details