Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1H3orf70
Entrez Gene ID
615631
Gene Name
chromosome 1 C3orf70 homolog
Gene Aliases
-
Chromosome Location
Chr.1: 81984026 - 82112760 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195030.1 NP_001181959.1 1 - 2 405 80
GenBank mRNA BC151738.1 - 1 - 2 347 80

View Full Product Details