The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037475.2 | NP_001032552.2 | 4 - 5 | 396 | 67 |
GenBank mRNA | BC108153.1 | - | 1 - 1 | 378 | 67 |
EE222978.1 | - | 4 - 5 | 397 | 67 | |
EV627197.1 | - | 4 - 5 | 384 | 67 |