Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRKAR1B
Entrez Gene ID
505370
Gene Name
protein kinase cAMP-dependent type I regulatory subunit beta
Gene Aliases
-
Chromosome Location
Chr.25: 41917430 - 41990189 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075201.1 NP_001068669.1 10 - 11 1035 103
GenBank mRNA BC118392.1 - 10 - 11 1035 103

View Full Product Details