Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1QL2
Entrez Gene ID
516982
Gene Name
complement C1q like 2
Gene Aliases
-
Chromosome Location
Chr.2: 70918684 - 70920915 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205836.1 NP_001192765.1 1 - 2 680 110

View Full Product Details