Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTGIR
Entrez Gene ID
520278
Gene Name
prostaglandin I2 receptor
Gene Aliases
-
Chromosome Location
Chr.18: 53748518 - 53755899 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015622.2 NP_001015622.2 2 - 2 921 106

View Full Product Details