Product Details

SNP ID
rs77111076
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241778280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATCAACAGCCCTGGGGACAAAG[A/G]GGACGGTAGTGTGTGGTGCCACTGG
Phenotype
MIM: 609186 MIM: 608237
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
D2HGDH PubMed Links

Gene Details

Gene
D2HGDH
Gene Name
D-2-hydroxyglutarate dehydrogenase
There are no transcripts associated with this gene.

Gene
GAL3ST2
Gene Name
galactose-3-O-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022134.2 Intron NP_071417.2

View Full Product Details