Product Details

SNP ID
rs74434110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112604876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTGGCTTTTGTGGCCAATAT[A/T]ACTGGACTCCCGTGGTATCAAGTTT
Phenotype
MIM: 608298
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC80 PubMed Links

Gene Details

Gene
CCDC80
Gene Name
coiled-coil domain containing 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199511.2 3625 UTR 3 NP_955805.1
NM_199512.2 3625 UTR 3 NP_955806.1
Gene
LINC01279
Gene Name
long intergenic non-protein coding RNA 1279
There are no transcripts associated with this gene.

View Full Product Details