Product Details

SNP ID
rs75575079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119825748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTCAATTTTCAATTAAAAAAT[A/G]TGAAAAATAAACCAGTAGATGACTG
Phenotype
MIM: 605004 MIM: 603065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSK3B PubMed Links

Gene Details

Gene
GSK3B
Gene Name
glycogen synthase kinase 3 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146156.1 2673 UTR 3 NP_001139628.1
NM_002093.3 2673 UTR 3 NP_002084.2
XM_006713610.2 2673 UTR 3 XP_006713673.1
XM_006713611.2 2673 UTR 3 XP_006713674.1
Gene
NR1I2
Gene Name
nuclear receptor subfamily 1 group I member 2
There are no transcripts associated with this gene.

View Full Product Details