Product Details

SNP ID
rs75961703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138499146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGATATATGTTCCTTTTAAAAG[C/T]ACCAAGTGTCAGATTATGTAAGGGT
Phenotype
MIM: 614310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP70 PubMed Links

Gene Details

Gene
CEP70
Gene Name
centrosomal protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288964.1 Intron NP_001275893.1
NM_001288965.1 Intron NP_001275894.1
NM_001288966.1 Intron NP_001275895.1
NM_001288967.1 Intron NP_001275896.1
NM_001320598.1 Intron NP_001307527.1
NM_001320599.1 Intron NP_001307528.1
NM_001320600.1 Intron NP_001307529.1
NM_024491.3 Intron NP_077817.2
XM_005247805.3 Intron XP_005247862.1
XM_011513209.2 Intron XP_011511511.1
XM_017007270.1 Intron XP_016862759.1
XM_017007271.1 Intron XP_016862760.1
XM_017007272.1 Intron XP_016862761.1
XM_017007273.1 Intron XP_016862762.1
XM_017007274.1 Intron XP_016862763.1
XM_017007275.1 Intron XP_016862764.1
XM_017007276.1 Intron XP_016862765.1
XM_017007277.1 Intron XP_016862766.1
XM_017007278.1 Intron XP_016862767.1
XM_017007279.1 Intron XP_016862768.1
XM_017007280.1 Intron XP_016862769.1
XM_017007281.1 Intron XP_016862770.1
XM_017007282.1 Intron XP_016862771.1
XM_017007283.1 Intron XP_016862772.1
XM_017007284.1 Intron XP_016862773.1

View Full Product Details