Product Details

SNP ID
rs76541465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:170460264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACAAGCTGTATTTATAACTTTG[A/G]ATATTAATTAAACAGTAACGTTTCT
Phenotype
MIM: 615720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928583 PubMed Links

Gene Details

Gene
LOC101928583
Gene Name
uncharacterized LOC101928583
There are no transcripts associated with this gene.

Gene
SLC7A14
Gene Name
solute carrier family 7 member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020949.2 9423 UTR 3 NP_066000.2

View Full Product Details