Product Details

SNP ID
rs75493366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:171600832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTATATGAGACTAAGATTTTCA[C/T]TTCATTAGCTCTCTCCTTCACCTTG
Phenotype
MIM: 602382
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLD1 PubMed Links

Gene Details

Gene
PLD1
Gene Name
phospholipase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130081.2 5553 UTR 3 NP_001123553.1
NM_002662.4 5553 UTR 3 NP_002653.1
XM_005247533.2 5553 UTR 3 XP_005247590.1
XM_005247534.2 5553 UTR 3 XP_005247591.1
XM_011512897.1 5553 Intron XP_011511199.1
XM_011512898.1 5553 Intron XP_011511200.1
XM_011512899.2 5553 UTR 3 XP_011511201.1
XM_017006623.1 5553 Intron XP_016862112.1
XM_017006624.1 5553 UTR 3 XP_016862113.1

View Full Product Details