Product Details

SNP ID
rs74736080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:171602222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCATCTTCTCTTATCTGGATCTG[A/T]TCCTCCTCGACACCTTGGGGGTTTC
Phenotype
MIM: 602382
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLD1 PubMed Links

Gene Details

Gene
PLD1
Gene Name
phospholipase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130081.2 4163 UTR 3 NP_001123553.1
NM_002662.4 4163 UTR 3 NP_002653.1
XM_005247533.2 4163 UTR 3 XP_005247590.1
XM_005247534.2 4163 UTR 3 XP_005247591.1
XM_011512897.1 4163 Intron XP_011511199.1
XM_011512898.1 4163 Intron XP_011511200.1
XM_011512899.2 4163 UTR 3 XP_011511201.1
XM_017006623.1 4163 Intron XP_016862112.1
XM_017006624.1 4163 UTR 3 XP_016862113.1

View Full Product Details