Product Details

SNP ID
rs77458308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9038115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCCAAGTGCATCAGGAAGTGA[A/T]GAAGAAGTTAGGATGAACAAATGGG
Phenotype
MIM: 609297
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SEMA5A PubMed Links

Gene Details

Gene
SEMA5A
Gene Name
semaphorin 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003966.2 8719 UTR 3 NP_003957.2
XM_006714506.2 8719 UTR 3 XP_006714569.1
XM_006714507.3 8719 UTR 3 XP_006714570.1
XM_011514155.2 8719 UTR 3 XP_011512457.1
XM_011514156.1 8719 UTR 3 XP_011512458.1
XM_011514157.1 8719 UTR 3 XP_011512459.1
XM_011514158.1 8719 UTR 3 XP_011512460.1
XM_011514159.2 8719 UTR 3 XP_011512461.1
XM_017010016.1 8719 UTR 3 XP_016865505.1

View Full Product Details