Product Details
- SNP ID
-
rs75456095
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:67182961 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAGAAAGAAAATGCCTATGGCTG[G/T]AATCCCACAGGAAAGCTTGACATCA
- Phenotype
-
MIM: 602226
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD180
PubMed Links
Gene Details
- Gene
- CD180
- Gene Name
- CD180 molecule
View Full Product Details