Product Details

SNP ID
rs74978501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73567999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTAGAAATCACAGTAAACTCAA[C/T]AGCTGAAGAGACAGAGTGGGGTGTG
Phenotype
MIM: 605787 MIM: 616194
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRA2 PubMed Links

Gene Details

Gene
ANKRA2
Gene Name
ankyrin repeat family A member 2
There are no transcripts associated with this gene.

Gene
UTP15
Gene Name
UTP15, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284430.1 Intron NP_001271359.1
NM_001284431.1 Intron NP_001271360.1
NM_032175.3 Intron NP_115551.2
XM_011543680.2 Intron XP_011541982.1

View Full Product Details