Product Details

SNP ID
rs75089353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:1611778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAGCAGCAGCTCGTCGTCCCTG[A/C]GTCACGGCGGCGGCGGCGGCGGCGG
Phenotype
MIM: 601090 MIM: 615976
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FOXC1 PubMed Links

Gene Details

Gene
FOXC1
Gene Name
forkhead box C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001453.2 1333 Missense Mutation AGT,CGT S445R NP_001444.2
Gene
FOXCUT
Gene Name
FOXC1 upstream transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details