Product Details

SNP ID
rs80286420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17291451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTTGTTAAACCTCTCTTACGTG[C/T]GTAGCCCTCATGAATCTTGATTTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM24 PubMed Links

Gene Details

Gene
RBM24
Gene Name
RNA binding motif protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143941.1 Intron NP_001137413.1
NM_001143942.1 Intron NP_001137414.1
NM_153020.2 Intron NP_694565.1
XM_005248932.3 Intron XP_005248989.1
XM_011514387.2 Intron XP_011512689.1
XM_011514388.2 Intron XP_011512690.1
XM_011514389.2 Intron XP_011512691.1

View Full Product Details