Product Details

SNP ID
rs78039629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35301608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCCTTCTTGCTCTCAGCAGGT[G/T]ACACAGCTAATCCAAAGATTCCCAC
Phenotype
MIM: 610094 MIM: 194549
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEF6 PubMed Links

Gene Details

Gene
DEF6
Gene Name
DEF6, guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022047.3 Intron NP_071330.3
Gene
ZNF76
Gene Name
zinc finger protein 76
There are no transcripts associated with this gene.

View Full Product Details