Product Details

SNP ID
rs76145865
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49551888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTGTTTCTCTTTAGGACCTAG[A/G]AACAGAAGTGAAGTTTGAAGTCTGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C6orf141 PubMed Links

Gene Details

Gene
C6orf141
Gene Name
chromosome 6 open reading frame 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145652.1 1245 UTR 3 NP_001139124.1
XM_005248850.3 1245 UTR 3 XP_005248907.1
XM_005248852.2 1245 Intron XP_005248909.1
XM_005248853.2 1245 Intron XP_005248910.1
XM_006714997.2 1245 UTR 3 XP_006715060.1
XM_011514303.2 1245 UTR 3 XP_011512605.1
XM_017010294.1 1245 UTR 3 XP_016865783.1
XM_017010295.1 1245 UTR 3 XP_016865784.1
XM_017010296.1 1245 UTR 3 XP_016865785.1
XM_017010297.1 1245 UTR 3 XP_016865786.1
XM_017010298.1 1245 UTR 3 XP_016865787.1

View Full Product Details