Product Details
- SNP ID
-
rs77514304
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:52672488 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTTTCCGGACTGTGTGTTGGGC[A/G]TCTCAGACTGAATACATTCTAAACA
- Phenotype
-
MIM: 616870
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC730101
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs543952570,rs578162238] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC730101
- Gene Name
- uncharacterized LOC730101
There are no transcripts associated with this gene.
- Gene
- TMEM14A
- Gene Name
- transmembrane protein 14A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014051.3 |
|
Intron |
|
|
NP_054770.1 |
View Full Product Details