Product Details

SNP ID
rs80097396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:7125296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGTAGGGTGAAACACTGGACAA[A/G]TTCTGCCAAGTATGGAATTGTCACC
Phenotype
MIM: 602209
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RREB1 PubMed Links

Gene Details

Gene
RREB1
Gene Name
ras responsive element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003698.3 Intron NP_001003698.1
NM_001003699.3 Intron NP_001003699.1
NM_001003700.1 Intron NP_001003700.1
NM_001168344.1 Intron NP_001161816.1
XM_006715157.3 Intron XP_006715220.1
XM_011514794.2 Intron XP_011513096.1
XM_011514795.2 Intron XP_011513097.1

View Full Product Details