Product Details

SNP ID
rs78160384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:158862071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCTCTGGGTCTCTTAGCGTGC[A/G]CCCTAGTTTGGTGTGTTGCCATGCA
Phenotype
MIM: 615462
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR60 PubMed Links

Gene Details

Gene
WDR60
Gene Name
WD repeat domain 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018051.4 Intron NP_060521.4
XM_005249549.1 Intron XP_005249606.1
XM_006716041.2 Intron XP_006716104.1
XM_011516367.1 Intron XP_011514669.1
XM_011516368.2 Intron XP_011514670.1
XM_011516369.2 Intron XP_011514671.1
XM_017012381.1 Intron XP_016867870.1
XM_017012382.1 Intron XP_016867871.1
XM_017012383.1 Intron XP_016867872.1
XM_017012384.1 Intron XP_016867873.1
XM_017012385.1 Intron XP_016867874.1
XM_017012386.1 Intron XP_016867875.1
XM_017012387.1 Intron XP_016867876.1
XM_017012388.1 Intron XP_016867877.1

View Full Product Details