Product Details

SNP ID
rs77632172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:22119300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTGAGCACTGACCTTATCCACA[C/T]GCTTCTAAATTCGGTGTATGCTCCA
Phenotype
MIM: 609527
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAPGEF5 PubMed Links

Gene Details

Gene
RAPGEF5
Gene Name
Rap guanine nucleotide exchange factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012294.3 7585 UTR 3 NP_036426.3
XM_005249914.2 7585 UTR 3 XP_005249971.1
XM_005249916.1 7585 UTR 3 XP_005249973.1
XM_011515652.2 7585 UTR 3 XP_011513954.1
XM_017012837.1 7585 UTR 3 XP_016868326.1

View Full Product Details