Product Details

SNP ID
rs77095806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30032961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGCCTGCATCATACTGGCCAG[A/G]TGGTTTTCATTAAGGATGTGCTATA
Phenotype
MIM: 614505 MIM: 608639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP14 PubMed Links

Gene Details

Gene
FKBP14
Gene Name
FK506 binding protein 14
There are no transcripts associated with this gene.

Gene
PLEKHA8
Gene Name
pleckstrin homology domain containing A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197026.1 Intron NP_001183955.1
NM_001197027.1 Intron NP_001183956.1
NM_032639.3 Intron NP_116028.1
XM_005249897.2 Intron XP_005249954.1
XM_011515591.2 Intron XP_011513893.1
XM_011515592.2 Intron XP_011513894.1
XM_011515594.1 Intron XP_011513896.1
XM_011515595.1 Intron XP_011513897.1
XM_011515596.2 Intron XP_011513898.1
XM_017012744.1 Intron XP_016868233.1
XM_017012745.1 Intron XP_016868234.1
XM_017012746.1 Intron XP_016868235.1
XM_017012747.1 Intron XP_016868236.1
XM_017012748.1 Intron XP_016868237.1
XM_017012749.1 Intron XP_016868238.1

View Full Product Details