Product Details

SNP ID
rs77373283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:50103587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGAGGCTCTCACAGAGCAGGACT[A/G]CCATTGCACATACTGTTTCCATTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf72 PubMed Links

Gene Details

Gene
C7orf72
Gene Name
chromosome 7 open reading frame 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161834.2 Intron NP_001155306.2
XM_011515052.1 Intron XP_011513354.1
XM_011515053.2 Intron XP_011513355.1

View Full Product Details