Product Details

SNP ID
rs75917357
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5060860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTAGATTTCTGAGCAGAGTACC[A/G]TCAGGGTTGCATTGAATGTGGGTGA
Phenotype
MIM: 608191
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RBAK PubMed Links

Gene Details

Gene
RBAK
Gene Name
RB associated KRAB zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204456.1 Intron NP_001191385.1
NM_021163.3 Intron NP_066986.1
Gene
RBAK-RBAKDN
Gene Name
RBAK-RBAKDN readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204513.1 Intron NP_001191442.1

View Full Product Details