Product Details

SNP ID
rs77440366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144050404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGCAGGTACTGTCACAAGGGCAT[A/G]ACAAAAAATCAGACATGGGTCACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2A5 PubMed Links

Gene Details

Gene
OR2A5
Gene Name
olfactory receptor family 2 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012365.1 Intron NP_036497.1

View Full Product Details