Product Details

SNP ID
rs75374828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:72895167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTGGAGCACTTCTTGCTCTTC[C/T]ACACCATCTCAATGACCAGGGTTGT
Phenotype
MIM: 615753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POM121 PubMed Links

Gene Details

Gene
POM121
Gene Name
POM121 transmembrane nucleoporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257190.2 Intron NP_001244119.1
NM_172020.4 Intron NP_742017.1
XM_005250726.3 Intron XP_005250783.1
XM_005250727.3 Intron XP_005250784.1
XM_006716196.3 Intron XP_006716259.1
XM_011516730.1 Intron XP_011515032.1
XM_017012853.1 Intron XP_016868342.1
XM_017012854.1 Intron XP_016868343.1

View Full Product Details