Product Details

SNP ID
rs77619496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95406204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTACCCAGATGTCCCCCGAGGAA[A/G]GATCAATAGATAAATTATCCACCAG
Phenotype
MIM: 602447 MIM: 602720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PON2 PubMed Links

Gene Details

Gene
PON2
Gene Name
paraoxonase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000305.2 817 Missense Mutation CCT,CTT P274L NP_000296.2
NM_001018161.1 817 Missense Mutation CCT,CTT P262L NP_001018171.1
XM_005250453.1 817 Missense Mutation CCT,CTT P206L XP_005250510.1
XM_017012357.1 817 Missense Mutation CCT,CTT P204L XP_016867846.1
XM_017012358.1 817 Missense Mutation CCT,CTT P188L XP_016867847.1
Gene
PON3
Gene Name
paraoxonase 3
There are no transcripts associated with this gene.

View Full Product Details