Product Details

SNP ID
rs77433080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101024489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGGAGAAGGGGGGTTGTGACCG[C/T]AACTGCTTCTGCCTCCTTCTCTTCC
Phenotype
MIM: 604609 MIM: 608424
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724094 PubMed Links

Gene Details

Gene
LOC102724094
Gene Name
uncharacterized LOC102724094
There are no transcripts associated with this gene.

Gene
MUC12
Gene Name
mucin 12, cell surface associated
There are no transcripts associated with this gene.

Gene
MUC17
Gene Name
mucin 17, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040105.1 Intron NP_001035194.1

View Full Product Details