Product Details

SNP ID
rs77855154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66378634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTTGGGTCCATCAAGGATGCCA[C/G]AATGCAGGCAGAAACCTGGCGTTGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00174 PubMed Links

Gene Details

Gene
LINC00174
Gene Name
long intergenic non-protein coding RNA 174
There are no transcripts associated with this gene.

View Full Product Details