Product Details

SNP ID
rs74199055
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101219050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGAAAGGTTACATTTTGAAAG[A/G]CATTATACACACTGCCCAGGAGTTT
Phenotype
MIM: 603066
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PLOD3 PubMed Links

Gene Details

Gene
PLOD3
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
There are no transcripts associated with this gene.

Gene
ZNHIT1
Gene Name
zinc finger HIT-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006349.2 Intron NP_006340.1
XM_011515739.2 Intron XP_011514041.1

View Full Product Details