Product Details

SNP ID
rs75697646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:132063168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGACTGTGCAACCCCACTGGAA[C/G]CTTGTGCCTGGGCTCTCTTGGACTC
Phenotype
MIM: 604109 MIM: 601658
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HHLA1 PubMed Links

Gene Details

Gene
HHLA1
Gene Name
HERV-H LTR-associating 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145095.1 2423 UTR 3 NP_001138567.1
Gene
OC90
Gene Name
otoconin 90
There are no transcripts associated with this gene.

View Full Product Details