Product Details

SNP ID
rs74339084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:57979746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATATCTTCTAAGATCTCAGCTA[A/G]GATCATTTGATACCCAAGACCTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01602 PubMed Links

Gene Details

Gene
LINC01602
Gene Name
long intergenic non-protein coding RNA 1602
There are no transcripts associated with this gene.

View Full Product Details