Product Details
- SNP ID
-
rs79457060
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:55107966 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATTCCAAGATGAATAACATATAT[A/C]TATTTCAAGGAATCTACTCTCTAAT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
XKR4
PubMed Links
Gene Details
- Gene
- XKR4
- Gene Name
- XK related 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052898.1 |
|
Intron |
|
|
NP_443130.1 |
View Full Product Details