Product Details

SNP ID
rs76945433
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33110914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCTCCTTTACTGTCCTGTGCCA[C/T]CTCTGCCCTCTCTGATCTTTCCCAA
Phenotype
MIM: 137060
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
B4GALT1 PubMed Links

Gene Details

Gene
B4GALT1
Gene Name
beta-1,4-galactosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001497.3 3836 UTR 3 NP_001488.2
XM_005251440.4 3836 UTR 3 XP_005251497.1

View Full Product Details