Product Details

SNP ID
rs74458889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34086861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGAGGGGGACAACTGGGAGAA[C/G]AAGTTAAAAAACTGGGGTGCAGGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCAF12 PubMed Links

Gene Details

Gene
DCAF12
Gene Name
DDB1 and CUL4 associated factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015397.3 3020 UTR 3 NP_056212.1
XM_011517843.1 3020 UTR 3 XP_011516145.1

View Full Product Details