Product Details

SNP ID
rs76429508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34655632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGTATGGGCAGCCAGGCAGGT[C/T]CGTGAAGCTGTGTTGTCCTGGAGTG
Phenotype
MIM: 604833 MIM: 606999 MIM: 600939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL27 PubMed Links

Gene Details

Gene
CCL27
Gene Name
C-C motif chemokine ligand 27
There are no transcripts associated with this gene.

Gene
GALT
Gene Name
galactose-1-phosphate uridylyltransferase
There are no transcripts associated with this gene.

Gene
IL11RA
Gene Name
interleukin 11 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142784.2 177 Missense Mutation TCC,TTC S43F NP_001136256.1
Gene
LOC730098
Gene Name
uncharacterized LOC730098
There are no transcripts associated with this gene.

View Full Product Details